siRNA Details
| Gene Symbol: WDR36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139281.2 | NP_644810.1 | 13, 14 | 1727 | ||
| GenBank mRNA | AF385437.1 | - | 13, 14 | 1727 | ||
| AK074202.1 | - | 7, 8 | 777 | |||
| AK095312.1 | - | 13, 14 | 1585 | |||
| AL832494.1 | - | 12, 13 | 1529 | |||
| BC133025.1 | - | 13, 14 | 1687 | 40147047 | ||
| BC136517.1 | - | 13, 14 | 1687 | 9020507 | ||
| CR749211.1 | - | 8, 9 | 927 | |||

WDR36
Genomic Map