siRNA Details
| Gene Symbol: C5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135586.1 | NP_001129058.1 | 2 | 599 | ||
| NM_001300894.1 | NP_001287823.1 | 2 | 599 | |||
| NM_152409.3 | NP_689622.2 | 2 | 412 | |||
| XM_005271889.3 | XP_005271946.1 | 2 | 1248 | |||
| XM_017009049.1 | XP_016864538.1 | 2 | 767 | |||
| XM_017009050.1 | XP_016864539.1 | 2 | 375 | |||
| GenBank mRNA | AJ437658.2 | - | 2 | 471 | ||
| AK094881.1 | - | 2 | 549 | |||
| BC053677.1 | - | 2 | 412 | 6166677 | ||
| BX648529.1 | - | 2 | 479 | |||

C5orf24
Genomic Map