siRNA Details
| Gene Symbol: C5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135586.1 | NP_001129058.1 | 2 | 421 | ||
| NM_001300894.1 | NP_001287823.1 | 2 | 421 | |||
| NM_152409.3 | NP_689622.2 | 2 | 234 | |||
| XM_005271889.3 | XP_005271946.1 | 2 | 1070 | |||
| XM_017009049.1 | XP_016864538.1 | 2 | 589 | |||
| XM_017009050.1 | XP_016864539.1 | 2 | 197 | |||
| GenBank mRNA | AJ437658.2 | - | 2 | 293 | ||
| AK094881.1 | - | 2 | 371 | |||
| BC053677.1 | - | 2 | 234 | 6166677 | ||
| BX648529.1 | - | 2 | 301 | |||
| DA433671.1 | - | 2 | 421 | |||
| DA760705.1 | - | 2 | 423 | |||

C5orf24
Genomic Map