siRNA Details
| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 736 | ||
| XM_006715336.3 | XP_006715399.1 | 4 | 777 | |||
| XM_011535447.1 | XP_011533749.1 | 4 | 776 | |||
| XM_017010264.1 | XP_016865753.1 | 4 | 555 | |||
| GenBank mRNA | AK098678.1 | - | 4 | 1233 | ||
| BC112253.1 | - | 4 | 725 | 8327721 | ||
| BC112255.1 | - | 4 | 725 | 8327723 | ||
| BX103697.1 | - | 2 | 312 | |||

IRAK1BP1
Genomic Map