siRNA Details
| Gene Symbol: TRIM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286633.1 | NP_001273562.1 | 5, 6 | 1083 | ||
| NM_138700.4 | NP_619645.1 | 4, 5 | 914 | |||
| XM_011514305.1 | XP_011512607.1 | 5, 6 | 1514 | |||
| XM_011514306.1 | XP_011512608.1 | 6, 7 | 1380 | |||
| GenBank mRNA | AF489517.1 | - | 4, 5 | 637 | ||
| BC060785.1 | - | 5, 6 | 1083 | 30340682 | ||

TRIM40
Genomic Map