siRNA Details
| Gene Symbol: OR2A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001659.1 | NP_001001659.1 | 1 | 817 | ||
| GenBank mRNA | BC136889.1 | - | 1 | 869 | 9020880 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2A14| Gene Symbol: OR2A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001659.1 | NP_001001659.1 | 1 | 817 | ||
| GenBank mRNA | BC136889.1 | - | 1 | 869 | 9020880 | |