siRNA Details
| Gene Symbol: ZNF786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152411.3 | NP_689624.2 | 3 | 368 | ||
| GenBank mRNA | AK095701.1 | - | 2 | 212 | ||
| AK297472.1 | - | 4 | 632 | |||
| AL834510.1 | - | 3 | 286 | |||
| BC109245.2 | - | 2 | 144 | |||
| BC128392.1 | - | 3 | 273 | 40007687 | ||
| BX647929.1 | - | 2 | 204 | |||
| DB453090.2 | - | 2 | 241 | |||

ZNF786
Genomic Map