siRNA Details
| Gene Symbol: ZNF786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152411.3 | NP_689624.2 | 3 | 390 | ||
| GenBank mRNA | AK095701.1 | - | 2 | 234 | ||
| AK297472.1 | - | 4 | 654 | |||
| AL834510.1 | - | 3 | 308 | |||
| BC109245.2 | - | 2 | 166 | |||
| BC128392.1 | - | 3 | 295 | 40007687 | ||
| BX647929.1 | - | 2 | 226 | |||
| DB453090.2 | - | 2 | 263 | |||

ZNF786
Genomic Map