siRNA Details
| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301821.1 | NP_001288750.1 | 12, 13 | 1319 | ||
| NM_001301822.1 | NP_001288751.1 | 11, 12 | 1234 | |||
| NM_130768.2 | NP_570124.1 | 12, 13 | 1346 | |||
| GenBank mRNA | AF461259.1 | - | 12, 13 | 1312 | ||
| AK093445.1 | - | 12, 13 | 1299 | |||
| BC028695.1 | - | 11, 12 | 2462 | |||
| BC034963.1 | - | 11, 12 | 2468 | 4824818 | ||
| BC068488.1 | - | 10, 11 | 2353 | |||
| BC126186.1 | - | 12, 13 | 1305 | 8991902 | ||
| BC126188.1 | - | 12, 13 | 1305 | 8991904 | ||
| BC144212.1 | - | 12, 13 | 1278 | 9052732 | ||

ASZ1
Genomic Map