siRNA Details
| Gene Symbol: CLDN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194284.2 | NP_919260.2 | 1 | 1671 | ||
| GenBank mRNA | AK123547.1 | - | 2 | 1296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLDN23| Gene Symbol: CLDN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194284.2 | NP_919260.2 | 1 | 1671 | ||
| GenBank mRNA | AK123547.1 | - | 2 | 1296 | ||