siRNA Details
| Gene Symbol: TMEM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286657.1 | NP_001273586.1 | 2 | 176 | ||
| NM_001286660.1 | NP_001273589.1 | 3 | 315 | |||
| NM_152417.2 | NP_689630.1 | 3 | 315 | |||
| XM_005251150.1 | XP_005251207.1 | 3 | 304 | |||
| XM_011517454.1 | XP_011515756.1 | 3 | 328 | |||
| XM_011517455.2 | XP_011515757.1 | 3 | 304 | |||
| GenBank mRNA | AK056932.1 | - | 3 | 234 | ||
| AK308247.1 | - | 3 | 218 | |||
| BC020835.1 | - | 3 | 274 | 4806471 | ||
| BC071636.1 | - | 3 | 275 | 5193338 | ||

TMEM68
Genomic Map