siRNA Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005225.2 | NP_005216.1 | 3 | 523 | ||
GenBank mRNA | BC050369.2 | - | 3 | 523 | 6025053 | |
BC058902.1 | - | 3 | 454 | 6055344 | ||
M96577.1 | - | 3 | 518 | |||
S49592.1 | - | 3 | 500 |

More Information
Other Species Targeted

Matching Taqman Gene Expression Assay:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
