siRNA Details
| Gene Symbol: SGCZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322880.1 | NP_001309809.1 | 2, 3 | 896 | ||
| NM_139167.3 | NP_631906.2 | 2, 3 | 896 | |||
| GenBank mRNA | AY028700.1 | - | 1, 2 | 197 | ||
| AY502063.1 | - | 2, 3 | 236 | |||
| BC069459.1 | - | 1, 2 | 197 | |||

SGCZ
Genomic Map