siRNA Details
| Gene Symbol: C9orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048265.1 | NP_001041730.1 | 2 | 233 | ||
| NM_144654.2 | NP_653255.1 | 2 | 233 | |||
| GenBank mRNA | AK024007.1 | - | 2 | 836 | ||
| AY927868.1 | - | 2 | 169 | |||
| BC021261.1 | - | 3 | 339 | 4136397 | ||
| DR003074.1 | - | 2 | 316 | |||

C9orf116
Genomic Map