siRNA Details
| Gene Symbol: C9orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048265.1 | NP_001041730.1 | 3 | 346 | ||
| NM_144654.2 | NP_653255.1 | 3 | 300 | |||
| GenBank mRNA | AK024007.1 | - | 3 | 903 | ||
| AY927868.1 | - | 3 | 282 | |||
| BC021261.1 | - | 4 | 406 | 4136397 | ||

C9orf116
Genomic Map