siRNA Details
| Gene Symbol: C9orf85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182505.3 | NP_872311.2 | 2 | 398 | ||
| XM_011518226.2 | XP_011516528.1 | 3 | 696 | |||
| GenBank mRNA | AK307186.1 | - | 2 | 211 | ||
| AK311206.1 | - | 3 | 397 | |||
| BC028019.1 | - | 2 | 244 | |||
| BC052375.1 | - | 2 | 237 | 6147448 | ||
| BQ431408.1 | - | 2 | 199 | |||

C9orf85
Genomic Map