siRNA Details
| Gene Symbol: ARID3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017363.1 | NP_001017363.1 | 4, 5 | 872 | ||
| XM_017014283.1 | XP_016869772.1 | 6, 7 | 3958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARID3C| Gene Symbol: ARID3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017363.1 | NP_001017363.1 | 4, 5 | 872 | ||
| XM_017014283.1 | XP_016869772.1 | 6, 7 | 3958 | |||