siRNA Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040410.2 | NP_001035500.1 | 2 | 1568 | ||
| NM_001040411.2 | NP_001035501.1 | 2 | 1457 | |||
| NM_001040412.2 | NP_001035502.1 | 3 | 1532 | |||
| NM_001287391.1 | NP_001274320.1 | 2 | 1489 | |||
| NM_203299.3 | NP_976044.2 | 2 | 1673 | |||
| GenBank mRNA | AK097595.1 | - | 2 | 1639 | ||
| BC045643.1 | - | 2 | 1652 | 5298387 | ||

C9orf131
Genomic Map