siRNA Details
| Gene Symbol: OR13F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004485.1 | NP_001004485.1 | 1 | 71 | ||
| GenBank mRNA | BC140739.1 | - | 1 | 160 | 9021607 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR13F1| Gene Symbol: OR13F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004485.1 | NP_001004485.1 | 1 | 71 | ||
| GenBank mRNA | BC140739.1 | - | 1 | 160 | 9021607 | |
Set Membership: |
|