siRNA Details
| Gene Symbol: SPANXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009609.3 | NP_001009609.2 | 2 | 425 | ||
| XM_017029264.1 | XP_016884753.1 | 2 | 545 | |||
| GenBank mRNA | BC062750.1 | - | 429 | 6618011 | ||
| DQ336128.1 | - | 2 | 341 | |||
| DQ336129.1 | - | 2 | 341 | |||

SPANXN3
Genomic Map