siRNA Details
| Gene Symbol: ADGRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153834.3 | NP_722576.3 | 6 | 5215 | ||
| XM_011531269.2 | XP_011529571.1 | 5 | 5221 | |||
| XM_011531271.2 | XP_011529573.1 | 3 | 4953 | |||
| GenBank mRNA | AY140954.1 | - | 2 | 4732 | ||
| AY882585.1 | - | 6 | 5215 | |||
| CR936801.1 | - | 3 | 4459 | |||
More Information
Set Membership: |
|

ADGRG4
Genomic Map