siRNA Details
| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207320.2 | NP_997203.1 | 1 | 255 | ||
| GenBank mRNA | AK098697.1 | - | 1 | 255 | ||
| BC137355.1 | - | 1 | 251 | 9021353 | ||
| BC137356.1 | - | 1 | 251 | 9021354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUD6A| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207320.2 | NP_997203.1 | 1 | 255 | ||
| GenBank mRNA | AK098697.1 | - | 1 | 255 | ||
| BC137355.1 | - | 1 | 251 | 9021353 | ||
| BC137356.1 | - | 1 | 251 | 9021354 | ||