siRNA Details
| Gene Symbol: GPR119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178471.2 | NP_848566.1 | 1 | 536 | ||
| GenBank mRNA | AY288416.1 | - | 1 | 446 | ||
| BC095502.1 | - | 1 | 470 | 30706606 | ||
| BC101166.1 | - | 1 | 446 | 40016011 | ||
| BC101167.1 | - | 1 | 446 | 40016012 | ||
| BC101168.2 | - | 1 | 446 | 40016013 | ||
| BC126179.1 | - | 1 | 536 | 8991895 | ||
| BC126181.1 | - | 1 | 536 | 8991897 | ||
More Information
Set Membership: |
|

GPR119
Genomic Map