siRNA Details
| Gene Symbol: DOCK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144658.3 | NP_653259.3 | 44 | 5019 | ||
| XM_005262368.3 | XP_005262425.1 | 45 | 5303 | |||
| XM_005262369.4 | XP_005262426.1 | 45 | 4997 | |||
| XM_011531276.2 | XP_011529578.1 | 45 | 4938 | |||
| XM_011531278.2 | XP_011529580.1 | 32 | 3424 | |||
| GenBank mRNA | AK054649.1 | - | 5 | 700 | ||
| AK056684.1 | - | 14 | 1533 | |||
| AK124735.1 | - | 39 | 4442 | |||
| AK125641.1 | - | 23 | 2531 | |||
| AY692226.1 | - | 44 | 5019 | |||
| BC047713.1 | - | 10 | 1110 | |||

DOCK11
Genomic Map