siRNA Details
| Gene Symbol: SPIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012968.2 | NP_001012986.2 | 1 | 795 | ||
| GenBank mRNA | AK126931.1 | - | 1 | 772 | ||
| AL833314.1 | - | 1 | 788 | |||
| BC043412.1 | - | 1 | 306 | 6156224 | ||
| BC112360.1 | - | 1 | 546 | 40032302 | ||

SPIN4
Genomic Map