siRNA Details
| Gene Symbol: RPS4Y2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039567.2 | NP_001034656.1 | 7 | 838 | ||
| GenBank mRNA | AF497481.2 | - | 7 | 838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS4Y2| Gene Symbol: RPS4Y2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039567.2 | NP_001034656.1 | 7 | 838 | ||
| GenBank mRNA | AF497481.2 | - | 7 | 838 | ||