siRNA Details
| Gene Symbol: KRTAP13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181599.2 | NP_853630.2 | 1 | 720 | ||
| GenBank mRNA | AJ457066.1 | - | 1 | 588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP13-1| Gene Symbol: KRTAP13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181599.2 | NP_853630.2 | 1 | 720 | ||
| GenBank mRNA | AJ457066.1 | - | 1 | 588 | ||