siRNA Details
| Gene Symbol: ASB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080874.3 | NP_543150.1 | 6 | 964 | ||
| XM_005262759.1 | XP_005262816.1 | 8 | 1393 | |||
| XM_011531616.2 | XP_011529918.1 | 6 | 1072 | |||
| XM_011531617.2 | XP_011529919.1 | 6 | 1655 | |||
| GenBank mRNA | BX647857.1 | - | 6 | 965 | ||

ASB5
Genomic Map