siRNA Details
| Gene Symbol: ASB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024708.3 | NP_078984.2 | 4, 5 | 986 | ||
| NM_198243.2 | NP_937886.1 | 4, 5 | 986 | |||
| GenBank mRNA | AF451994.1 | - | 4, 5 | 906 | ||
| AK026204.1 | - | 4, 5 | 906 | |||
| AK222958.1 | - | 4, 5 | 906 | |||
| AK222974.1 | - | 4, 5 | 906 | |||
| AK289860.1 | - | 4, 5 | 939 | |||
| BC063581.1 | - | 4, 5 | 927 | 4370796 | ||

ASB7
Genomic Map