siRNA Details
| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199629.1 | NP_001186558.1 | 2 | 380 | ||
| NM_002475.4 | NP_002466.1 | 2 | 316 | |||
| GenBank mRNA | AK304594.1 | - | 2 | 149 | ||
| AK310996.1 | - | 2 | 146 | |||
| BC012425.1 | - | 1 | 104 | 3862144 | ||
| BC014400.1 | - | 2 | 380 | 3503030 | ||
| BU166567.1 | - | 203 | ||||
| M31211.1 | - | 1 | 84 | |||
| X16434.1 | - | 1 | 69 | |||

MYL6B
Genomic Map