siRNA Details
Gene Symbol: GPR183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004951.4 | NP_004942.1 | 2 | 428 | ||
XM_017020405.1 | XP_016875894.1 | 2 | 371 | |||
GenBank mRNA | AK223390.1 | - | 2 | 332 | ||
AK292091.1 | - | 2 | 352 | |||
AK313443.1 | - | 2 | 428 | |||
AW402166.1 | - | 2 | 380 | |||
BC020752.1 | - | 2 | 332 | 4722289 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
