siRNA Details
| Gene Symbol: GATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080473.4 | NP_536721.1 | 7 | 2157 | ||
| XM_006723699.3 | XP_006723762.1 | 7 | 2177 | |||
| GenBank mRNA | BC047790.1 | - | 7 | 2157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GATA5| Gene Symbol: GATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080473.4 | NP_536721.1 | 7 | 2157 | ||
| XM_006723699.3 | XP_006723762.1 | 7 | 2177 | |||
| GenBank mRNA | BC047790.1 | - | 7 | 2157 | ||