siRNA Details
| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080627.3 | NP_542194.2 | 13, 14 | 3819 | ||
| NM_199181.2 | NP_954650.2 | 13, 14 | 3092 | |||
| GenBank mRNA | AK126630.1 | - | 13, 14 | 3092 | ||
| AK131491.1 | - | 9, 10 | 2319 | |||

SOGA1
Genomic Map