siRNA Details
| Gene Symbol: TLDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304783.1 | NP_001291712.1 | 3 | 307 | ||
| NM_080628.2 | NP_542195.1 | 3 | 307 | |||
| XM_017027672.1 | XP_016883161.1 | 3 | 297 | |||
| XM_017027673.1 | XP_016883162.1 | 4 | 422 | |||
| XM_017027674.1 | XP_016883163.1 | 2 | 147 | |||
| GenBank mRNA | AK123544.1 | - | 3 | 261 | ||
| BC127688.1 | - | 3 | 307 | 40127440 | ||
| BC130646.1 | - | 3 | 296 | 40146677 | ||
| BC130648.1 | - | 3 | 296 | 40146679 | ||
| BY994247.1 | - | 3 | 295 | |||

TLDC2
Genomic Map