siRNA Details
| Gene Symbol: ANKRD60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304369.1 | NP_001291298.1 | 2 | 512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD60| Gene Symbol: ANKRD60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304369.1 | NP_001291298.1 | 2 | 512 | ||