siRNA Details
| Gene Symbol: TMEM37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183240.2 | NP_899063.2 | 2 | 1242 | ||
| XM_005263598.4 | XP_005263655.1 | 2 | 1542 | |||
| XM_006712300.3 | XP_006712363.1 | 2 | 1366 | |||
| XM_011510659.2 | XP_011508961.1 | 2 | 1259 | |||
| GenBank mRNA | AK026464.1 | - | 1 | 690 | ||
| BC046362.1 | - | 1 | 1368 | 5221396 | ||

TMEM37
Genomic Map