siRNA Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080748.2 | NP_542786.1 | 3 | 324 | ||
| XM_017027678.1 | XP_016883167.1 | 3 | 319 | |||
| GenBank mRNA | AK000548.1 | - | 3 | 229 | ||
| AM397244.1 | - | 3 | 223 | |||
| AM397245.1 | - | 3 | 229 | |||
| AM397246.1 | - | 2 | 304 | |||
| BC008488.1 | - | 340 | 4133274 | |||
| BC105281.1 | - | 3 | 233 | 6739248 | ||
| GU727625.1 | - | 3 | 233 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164217, NM_001164216, NM_025946, CJ221444, AA058111, and others.

ROMO1
Genomic Map