siRNA Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040022.1 | NP_001035111.1 | 6 | 1468 | ||
| NM_001040023.1 | NP_001035112.1 | 5 | 1376 | |||
| NM_080792.2 | NP_542970.1 | 6 | 1135 | |||
| XM_005260669.3 | XP_005260726.1 | 6 | 1518 | |||
| XM_005260670.3 | XP_005260727.1 | 6 | 1134 | |||
| XM_006723545.3 | XP_006723608.1 | 3 | 705 | |||
| XM_011529173.2 | XP_011527475.1 | 5 | 1123 | |||
| GenBank mRNA | AB023430.1 | - | 6 | 1135 | ||
| AK290776.1 | - | 6 | 1241 | |||
| AK298243.1 | - | 6 | 1086 | |||
| AK312521.1 | - | 5 | 1189 | |||
| BC026692.1 | - | 6 | 1140 | 3916836 | ||
| BC033092.1 | - | 5 | 1128 | 4589298 | ||
| BC038510.2 | - | 5 | 1373 | 5261789 | ||
| BC075849.1 | - | 6 | 1314 | 4589732 | ||
| D86043.1 | - | 5 | 1107 | |||
| U06701.1 | - | 4 | 1035 | |||
| Y10375.1 | - | 5 | 1117 | |||
| Y11047.1 | - | 4 | 883 | |||

SIRPA
Genomic Map