siRNA Details
| Gene Symbol: PABPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080832.2 | NP_543022.1 | 2 | 1172 | ||
| GenBank mRNA | AK290105.1 | - | 1 | 737 | ||
| AK297327.1 | - | 3 | 845 | |||
| AL122118.1 | - | 1 | 687 | |||
| BC063113.1 | - | 2 | 950 | 6452933 | ||

PABPC5
Genomic Map