siRNA Details
| Gene Symbol: HOXB-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033201.2 | 3 | 500 | |||
| NR_033204.2 | 3 | 403 | ||||
| GenBank mRNA | BC010732.1 | - | 3 | 358 | 3896690 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXB-AS3| Gene Symbol: HOXB-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033201.2 | 3 | 500 | |||
| NR_033204.2 | 3 | 403 | ||||
| GenBank mRNA | BC010732.1 | - | 3 | 358 | 3896690 | |