siRNA Details
| Gene Symbol: RBBP8NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080833.2 | NP_543023.2 | 12 | 1915 | ||
| GenBank mRNA | AK074919.1 | - | 12 | 1875 | ||
| AK128718.1 | - | 2 | 1795 | |||
| AK290299.1 | - | 12 | 1888 | |||
| BC006450.1 | - | 5 | 1120 | |||
| BC033142.1 | - | 5 | 1120 | |||
| BC137326.1 | - | 11 | 1803 | 9021324 | ||
| BC137327.1 | - | 11 | 1803 | 9021325 | ||

RBBP8NL
Genomic Map