siRNA Details
| Gene Symbol: CNBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207076.2 | NP_001194005.1 | 6 | 877 | ||
| NM_080834.3 | NP_543024.2 | 6 | 877 | |||
| NR_130714.1 | 5 | 720 | ||||
| XM_005260295.2 | XP_005260352.1 | 6 | 851 | |||
| XM_011528590.2 | XP_011526892.1 | 6 | 837 | |||
| XM_011528592.1 | XP_011526894.1 | 5 | 707 | |||
| XM_011528593.2 | XP_011526895.1 | 5 | 570 | |||
| XM_011528598.1 | XP_011526900.1 | 2 | 207 | |||
| XM_017027683.1 | XP_016883172.1 | 6 | 1055 | |||
| GenBank mRNA | AK057454.1 | - | 6 | 851 | ||
| BC114634.1 | - | 5 | 678 | |||
| BC114945.1 | - | 6 | 835 | 40084218 | ||

CNBD2
Genomic Map