siRNA Details
Gene Symbol: LINC01133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038849.1 | 2 | 216 | |||
GenBank mRNA | AF088076.1 | - | 2 | 156 | ||
CB119247.1 | - | 2 | 216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038849.1 | 2 | 216 | |||
GenBank mRNA | AF088076.1 | - | 2 | 156 | ||
CB119247.1 | - | 2 | 216 |