siRNA Details
| Gene Symbol: CNBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080834.3 | NP_543024.2 | 11 | 1514 | ||
| XM_005260295.2 | XP_005260352.1 | 11 | 1500 | |||
| XM_011528590.2 | XP_011526892.1 | 11 | 1486 | |||
| XM_011528592.1 | XP_011526894.1 | 10 | 1356 | |||
| XM_011528593.2 | XP_011526895.1 | 10 | 1219 | |||
| XM_011528598.1 | XP_011526900.1 | 7 | 856 | |||
| XM_017027683.1 | XP_016883172.1 | 11 | 1704 | |||
| GenBank mRNA | AK057454.1 | - | 11 | 1488 | ||

CNBD2
Genomic Map