siRNA Details
| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080836.3 | NP_543026.2 | 3 | 1423 | ||
| GenBank mRNA | AF195026.1 | - | 2 | 1024 | ||
| AK314728.1 | - | 3 | 1275 | |||
| AL040176.1 | - | 1 | 132 | |||
| BC017340.2 | - | 2 | 1006 | |||
| BC111573.1 | - | 2 | 563 | |||
| BC140883.1 | - | 2 | 1124 | 9021751 | ||
| BC140884.1 | - | 2 | 1124 | 9021752 | ||
| GQ281297.1 | - | 3 | 1423 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183262, BC047277, BC138904, BC138905, AK006778.

STK35
Genomic Map