siRNA Details
| Gene Symbol: R3HDML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178491.3 | NP_848586.1 | 1 | 193 | ||
| GenBank mRNA | BC107048.2 | - | 1 | 193 | 40003509 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3HDML| Gene Symbol: R3HDML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178491.3 | NP_848586.1 | 1 | 193 | ||
| GenBank mRNA | BC107048.2 | - | 1 | 193 | 40003509 | |