siRNA Details
| Gene Symbol: DCANP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130848.2 | NP_570900.1 | 1 | 658 | ||
| GenBank mRNA | AB074498.1 | - | 1 | 658 | ||
| BC069608.1 | - | 1 | 502 | 7262210 | ||
| BC069610.1 | - | 1 | 502 | 7262222 | ||
| BC074831.2 | - | 1 | 444 | 30915544 | ||
| BC074832.2 | - | 1 | 444 | 30915326 | ||
| BC117156.1 | - | 1 | 597 | |||
| BC117158.1 | - | 1 | 597 | |||

DCANP1
Genomic Map