siRNA Details
| Gene Symbol: RAB40A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080879.2 | NP_543155.2 | 3 | 1246 | ||
| XM_005262083.4 | XP_005262140.1 | 2 | 1723 | |||
| XM_011530872.2 | XP_011529174.1 | 2 | 1720 | |||
| GenBank mRNA | AF132748.1 | - | 1 | 969 | ||
| AF422143.1 | - | 3 | 1249 | |||
| AK056859.1 | - | 1 | 3023 | |||

RAB40A
Genomic Map