siRNA Details
| Gene Symbol: FGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152429.4 | NP_689642.3 | 2 | 1221 | ||
| GenBank mRNA | AK075410.1 | - | 2 | 1204 | ||
| BC025966.1 | - | 2 | 1190 | 4895171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFBP3| Gene Symbol: FGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152429.4 | NP_689642.3 | 2 | 1221 | ||
| GenBank mRNA | AK075410.1 | - | 2 | 1204 | ||
| BC025966.1 | - | 2 | 1190 | 4895171 | ||