siRNA Details
| Gene Symbol: KDELC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153705.4 | NP_714916.3 | 3 | 686 | ||
| XM_011542621.2 | XP_011540923.1 | 3 | 659 | |||
| XM_011542623.2 | XP_011540925.1 | 3 | 653 | |||
| GenBank mRNA | AF533708.1 | - | 3 | 645 | ||
| AK125519.1 | - | 2 | 468 | |||
| AK313534.1 | - | 3 | 548 | |||
| AY358616.1 | - | 2 | 755 | |||
| BC036526.2 | - | 3 | 640 | 5266665 | ||

KDELC2
Genomic Map